Assay Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027295.2 | NP_081571.1 | 3 - 4 | 433 | 120 | |
| GenBank mRNA | AB232624.1 | - | 3 - 4 | 257 | 120 | |
| AK012286.1 | - | 3 - 4 | 436 | 120 | ||
| AK159493.1 | - | 3 - 4 | 434 | 120 | ||
| AK161391.1 | - | 3 - 4 | 314 | 120 | ||
| BC004580.1 | - | 3 - 4 | 433 | 3587008 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161391.1 transcripts.
Set Membership: |
|

Rab28
Genomic Map