Assay Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC029122.1 | - | 2 - 3 | 204 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC029122.1 transcripts.
Set Membership: |
|

Zbtb49
Genomic Map