Assay Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029162.2 | NP_083438.1 | 5 - 6 | 1470 | 93 | |
| GenBank mRNA | AK015819.1 | - | 3 - 4 | 382 | 93 | |
| AK043044.1 | - | 5 - 6 | 1465 | 93 | ||
| AK054488.1 | - | 2 - 3 | 162 | 93 | ||
| AK140546.1 | - | 5 - 6 | 1465 | 93 | ||
| BC028988.1 | - | 4 - 5 | 1407 | 5376301 | 93 | |
| BC029122.1 | - | 3 - 4 | 283 | 93 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK148068.1 | - | 1476 | Chr. - - [-] on Build | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC029122.1 transcripts.
Set Membership: |
|

Zbtb49
Genomic Map