Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113423.2 | NP_001106894.1 | 6 - 7 | 1625 | 73 | |
| GenBank mRNA | AK129365.1 | - | 6 - 7 | 1614 | 73 | |
| BC076573.1 | - | 6 - 7 | 1558 | 73 | ||
More Information
Set Membership: |
|

Slain2
Genomic Map