Assay Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199032.2 | NP_950197.1 | 20 - 21 | 2850 | 65 | |
| GenBank mRNA | AK076392.1 | - | 13 - 14 | 1717 | 65 | |
| AK137244.1 | - | 21 - 22 | 3077 | 65 | ||
| AK172988.1 | - | 20 - 21 | 2761 | 65 | ||
| BC062951.1 | - | 20 - 21 | 2851 | 6852711 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep135
Genomic Map