Assay Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199032.2 | NP_950197.1 | 1 - 2 | 156 | 63 | |
| GenBank mRNA | AK137244.1 | - | 2 - 3 | 383 | 63 | |
| AK172988.1 | - | 1 - 2 | 67 | 63 | ||
| BC062951.1 | - | 1 - 2 | 157 | 6852711 | 63 | |
More Information
Set Membership: |
|

Cep135
Genomic Map