| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500006471.mm.1 |
TC0500000724.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006471.mm.1 |
NM_199032.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006471.mm.1 |
NM_199032.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006468.mm.1 |
TC0500000724.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006468.mm.1 |
NM_199032.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006468.mm.1 |
NM_199032.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006469.mm.1 |
TC0500000724.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006469.mm.1 |
NM_199032.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006469.mm.1 |
NM_199032.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006470.mm.1 |
TC0500000724.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500006470.mm.1 |
NM_199032.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500006470.mm.1 |
NM_199032.2 |
Refseq |