Assay Details
| Gene Symbol: Tmprss11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033233.2 | NP_001028405.1 | 7 - 8 | 1001 | 62 | |
| GenBank mRNA | AK142820.1 | - | 7 - 8 | 1002 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11a
Genomic Map