Assay Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133862.1 | NP_598623.1 | 4 - 5 | 412 | 107 | |
| GenBank mRNA | AK013887.1 | - | 4 - 5 | 451 | 108 | |
| AK149375.1 | - | 4 - 5 | 448 | 107 | ||
| AK149448.1 | - | 4 - 5 | 449 | 107 | ||
| BC019506.1 | - | 4 - 5 | 412 | 4193631 | 107 | |
| BC019828.1 | - | 4 - 5 | 422 | 5123696 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgg
Genomic Map