Assay Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133862.1 | NP_598623.1 | 6 - 7 | 672 | 105 | |
| GenBank mRNA | AF413206.1 | - | 2 - 3 | 153 | 105 | |
| AK149375.1 | - | 6 - 7 | 708 | 105 | ||
| AK149448.1 | - | 6 - 7 | 709 | 105 | ||
| BC019506.1 | - | 6 - 7 | 672 | 4193631 | 105 | |
| BC019828.1 | - | 6 - 7 | 682 | 5123696 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgg
Genomic Map