Assay Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 1 - 2 | 239 | 75 | |
| GenBank mRNA | AK052871.1 | - | 1 - 2 | 137 | 75 | |
| AK078287.1 | - | 1 - 2 | 171 | 75 | ||
| AK087261.1 | - | 1 - 2 | 240 | 75 | ||
| AK155930.1 | - | 1 - 2 | 175 | 75 | ||
| AY553221.1 | - | 1 - 2 | 115 | 75 | ||
| BC027369.1 | - | 1 - 2 | 218 | 75 | ||
| BC106095.1 | - | 1 - 2 | 159 | 75 | ||
| BC127163.1 | - | 1 - 2 | 105 | 75 | ||
| BC152540.1 | - | 1 - 2 | 107 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052871.1 transcripts.
Set Membership: |
|

Ddx58
Genomic Map