Assay Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 3 - 4 | 548 | 107 | |
| GenBank mRNA | AK078287.1 | - | 3 - 4 | 480 | 107 | |
| AK087261.1 | - | 3 - 4 | 549 | 107 | ||
| AK128929.1 | - | 3 - 4 | 483 | 107 | ||
| AK155930.1 | - | 3 - 4 | 484 | 107 | ||
| AY553221.1 | - | 3 - 4 | 424 | 107 | ||
| BC027369.1 | - | 3 - 4 | 527 | 107 | ||
| BC106095.1 | - | 3 - 4 | 468 | 107 | ||
| BC127163.1 | - | 3 - 4 | 414 | 107 | ||
| BC152540.1 | - | 3 - 4 | 416 | 107 | ||
More Information
Set Membership: |
|

Ddx58
Genomic Map