Assay Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 6 - 7 | 927 | 141 | |
| GenBank mRNA | AK052595.1 | - | 2 - 3 | 181 | 141 | |
| AK052871.1 | - | 6 - 7 | 912 | 141 | ||
| AK078287.1 | - | 6 - 7 | 859 | 141 | ||
| AK128929.1 | - | 6 - 7 | 862 | 141 | ||
| AK155930.1 | - | 6 - 7 | 863 | 141 | ||
| AY553221.1 | - | 6 - 7 | 803 | 141 | ||
| BC027369.1 | - | 6 - 7 | 906 | 141 | ||
| BC106095.1 | - | 6 - 7 | 847 | 141 | ||
| BC127163.1 | - | 6 - 7 | 793 | 141 | ||
| BC152540.1 | - | 6 - 7 | 795 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx58
Genomic Map