Assay Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 8 - 9 | 1341 | 68 | |
| GenBank mRNA | AK052595.1 | - | 4 - 5 | 595 | 68 | |
| AK128929.1 | - | 8 - 9 | 1276 | 68 | ||
| AK155930.1 | - | 8 - 9 | 1277 | 68 | ||
| AY553221.1 | - | 8 - 9 | 1217 | 68 | ||
| BC027369.1 | - | 8 - 9 | 1320 | 68 | ||
| BC106095.1 | - | 8 - 9 | 1261 | 68 | ||
| BC127163.1 | - | 8 - 9 | 1207 | 68 | ||
| BC152540.1 | - | 8 - 9 | 1209 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx58
Genomic Map