Assay Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 9 - 10 | 1504 | 60 | |
| GenBank mRNA | AK052595.1 | - | 5 - 6 | 758 | 60 | |
| AK128929.1 | - | 9 - 10 | 1439 | 60 | ||
| AK155930.1 | - | 9 - 10 | 1440 | 60 | ||
| BC027369.1 | - | 9 - 10 | 1483 | 60 | ||
| BC106095.1 | - | 9 - 10 | 1424 | 60 | ||
| BC127163.1 | - | 9 - 10 | 1370 | 60 | ||
| BC152540.1 | - | 9 - 10 | 1372 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx58
Genomic Map