Assay Details
| Gene Symbol: Topors | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134097.3 | NP_598858.2 | 1 - 2 | 171 | 75 | |
| GenBank mRNA | AB072395.1 | - | 1 - 2 | 128 | 75 | |
| AB104865.1 | - | 1 - 2 | 156 | 75 | ||
| AK044564.1 | - | 1 - 2 | 149 | 75 | ||
| AK134075.1 | - | 1 - 2 | 167 | 75 | ||
| AK140250.1 | - | 1 - 2 | 172 | 75 | ||
| AK143025.1 | - | 1 - 2 | 152 | 75 | ||
| AK153743.1 | - | 1 - 2 | 158 | 75 | ||
| BC019421.1 | - | 1 - 2 | 129 | 75 | ||
| BC037141.1 | - | 1 - 2 | 149 | 75 | ||
| BC040797.1 | - | 1 - 2 | 159 | 3492178 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB072395.1, AB104865.1, AK044564.1, AK134075.1, AK140250.1, AK143025.1, AK153743.1, BC019421.1, BC037141.1, BC040797.1, NM_134097.3 transcripts.
Set Membership: |
|

Topors
Genomic Map