Assay Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290425.1 | NP_001277354.1 | 9 - 10 | 998 | 72 | |
| NM_152825.2 | NP_690038.1 | 9 - 10 | 998 | 72 | ||
| GenBank mRNA | AK019756.1 | - | 10 - 11 | 1113 | 72 | |
| AK088073.1 | - | 9 - 10 | 981 | 72 | ||
| BC027768.1 | - | 9 - 10 | 981 | 5354502 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp45
Genomic Map