Assay Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290425.1 | NP_001277354.1 | 13 - 14 | 1888 | 70 | |
| NM_152825.2 | NP_690038.1 | 14 - 15 | 2032 | 70 | ||
| GenBank mRNA | AK019756.1 | - | 15 - 16 | 2147 | 70 | |
| AK036903.1 | - | 1 - 2 | 448 | 70 | ||
| AK088073.1 | - | 13 - 14 | 1871 | 70 | ||
| BC027768.1 | - | 14 - 15 | 2015 | 5354502 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp45
Genomic Map