Assay Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290425.1 | NP_001277354.1 | 4 - 5 | 434 | 92 | |
| NM_152825.2 | NP_690038.1 | 4 - 5 | 434 | 92 | ||
| GenBank mRNA | AK019756.1 | - | 5 - 6 | 549 | 92 | |
| AK040660.1 | - | 4 - 5 | 435 | 92 | ||
| AK088073.1 | - | 4 - 5 | 417 | 92 | ||
| AK162825.1 | - | 4 - 5 | 435 | 92 | ||
| BB213271.2 | - | 4 - 5 | 446 | 92 | ||
| BC027768.1 | - | 4 - 5 | 417 | 5354502 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp45
Genomic Map