Assay Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290425.1 | NP_001277354.1 | 6 - 7 | 676 | 125 | |
| NM_152825.2 | NP_690038.1 | 6 - 7 | 676 | 125 | ||
| GenBank mRNA | AK019756.1 | - | 7 - 8 | 791 | 125 | |
| AK088073.1 | - | 6 - 7 | 659 | 125 | ||
| BC027768.1 | - | 6 - 7 | 659 | 5354502 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp45
Genomic Map