Assay Details
| Gene Symbol: Ndufaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026742.4 | NP_081018.1 | 1 - 2 | 218 | 107 | |
| GenBank mRNA | AK003553.2 | - | 1 - 2 | 173 | 107 | |
| AK034728.1 | - | 1 - 2 | 201 | 107 | ||
| AK160341.1 | - | 1 - 2 | 214 | 107 | ||
| BC028764.1 | - | 1 - 2 | 187 | 1195846 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufaf4
Genomic Map