Assay Details
| Gene Symbol: Ankrd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030781.2 | 1 - 2 | 225 | 79 | ||
| GenBank mRNA | AK014768.1 | - | 1 - 2 | 225 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd10
Genomic Map