Assay Details
| Gene Symbol: Ankrd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281975.1 | NP_001268904.1 | 4 - 5 | 750 | 57 | |
| GenBank mRNA | AK081754.1 | - | 4 - 5 | 748 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd10
Genomic Map