Assay Details
| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133962.3 | NP_598723.3 | 17 - 18 | 2614 | 106 | |
| GenBank mRNA | AK147923.1 | - | 17 - 18 | 2587 | 106 | |
| AK172963.1 | - | 11 - 12 | 1717 | 106 | ||
| BC034512.1 | - | 6 - 7 | 106 | |||
| BC060639.1 | - | 17 - 18 | 2586 | 6849875 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef18
Genomic Map