Assay Details
| Gene Symbol: Fgfr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163215.2 | NP_001156687.1 | 3 - 4 | 709 | 66 | |
| NM_001163217.2 | NP_001156689.1 | 2 - 3 | 478 | 66 | ||
| NM_001205270.1 | NP_001192199.1 | 3 - 4 | 683 | 66 | ||
| NM_008010.5 | NP_032036.2 | 3 - 4 | 683 | 66 | ||
| GenBank mRNA | AK140217.1 | - | 2 - 3 | 600 | 66 | |
| AK143592.1 | - | 3 - 4 | 616 | 66 | ||
| AK163495.1 | - | 1 - 2 | 262 | 66 | ||
| AK163547.1 | - | 2 - 3 | 600 | 66 | ||
| BC053056.1 | - | 3 - 4 | 593 | 5708838 | 66 | |
| BY732245.1 | - | 3 - 4 | 647 | 66 | ||
| CB247234.1 | - | 3 - 4 | 554 | 66 | ||
| M81342.1 | - | 3 - 4 | 601 | 66 | ||
| S56291.1 | - | 2 - 3 | 428 | 66 | ||
| X58255.1 | - | 3 - 4 | 587 | 66 | ||
More Information
Set Membership: |
|

Fgfr3
Genomic Map