Assay Details
| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027276.2 | NP_081552.2 | 15 - 16 | 1485 | 111 | |
| GenBank mRNA | AK013213.1 | - | 4 - 5 | 419 | 111 | |
| AK147938.1 | - | 15 - 16 | 1500 | 111 | ||
| AK167965.1 | - | 15 - 16 | 1500 | 111 | ||
| AK168978.1 | - | 15 - 16 | 1472 | 111 | ||
| AK201828.1 | - | 3 - 4 | 204 | 111 | ||
| BC026606.1 | - | 15 - 16 | 1485 | 4976424 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc16
Genomic Map