Assay Details
| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027276.2 | NP_081552.2 | 1 - 2 | 164 | 78 | |
| GenBank mRNA | AK147938.1 | - | 1 - 2 | 179 | 78 | |
| AK167965.1 | - | 1 - 2 | 179 | 78 | ||
| AK168978.1 | - | 1 - 2 | 151 | 78 | ||
| BC026606.1 | - | 1 - 2 | 164 | 4976424 | 78 | |
More Information
Set Membership: |
|

Cdc16
Genomic Map