Assay Details
| Gene Symbol: Mxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010753.2 | NP_034883.2 | 4 - 5 | 442 | 81 | |
| GenBank mRNA | AK007824.1 | - | 4 - 5 | 409 | 81 | |
| AK131132.1 | - | 4 - 5 | 412 | 81 | ||
| AK134169.1 | - | 3 - 4 | 423 | 81 | ||
| BC011303.1 | - | 4 - 5 | 442 | 3490469 | 81 | |
| U32395.1 | - | 4 - 5 | 305 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxd4
Genomic Map