Assay Details
| Gene Symbol: Poln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289803.1 | NP_001276732.1 | 17 - 18 | 2081 | 144 | |
| NM_001289804.1 | NP_001276733.1 | 19 - 20 | 2184 | 144 | ||
| NM_181857.4 | NP_862905.1 | 18 - 19 | 2135 | 144 | ||
| GenBank mRNA | AK133063.1 | - | 19 - 20 | 2104 | 144 | |
| AY135562.3 | - | 18 - 19 | 2057 | 144 | ||
| BC141101.1 | - | 18 - 19 | 2035 | 9055713 | 144 | |
| BC145378.1 | - | 17 - 18 | 1981 | 9053936 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poln
Genomic Map