Assay Details
| Gene Symbol: Poln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289803.1 | NP_001276732.1 | 20 - 21 | 2405 | 67 | |
| NM_001289804.1 | NP_001276733.1 | 21 - 22 | 2376 | 67 | ||
| NM_181857.4 | NP_862905.1 | 21 - 22 | 2459 | 67 | ||
| GenBank mRNA | AK133063.1 | - | 21 - 22 | 2296 | 67 | |
| AY135562.3 | - | 21 - 22 | 2381 | 67 | ||
| BC141101.1 | - | 21 - 22 | 2359 | 9055713 | 67 | |
| BC145378.1 | - | 20 - 21 | 2305 | 9053936 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poln
Genomic Map