Assay Details
| Gene Symbol: Poln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289803.1 | NP_001276732.1 | 9 - 10 | 1454 | 102 | |
| NM_001289804.1 | NP_001276733.1 | 10 - 11 | 1503 | 102 | ||
| NM_181857.4 | NP_862905.1 | 9 - 10 | 1454 | 102 | ||
| GenBank mRNA | AK133063.1 | - | 10 - 11 | 1423 | 102 | |
| AY135562.3 | - | 9 - 10 | 1376 | 102 | ||
| BC141101.1 | - | 9 - 10 | 1354 | 9055713 | 102 | |
| BC145378.1 | - | 9 - 10 | 1354 | 9053936 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poln
Genomic Map