| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500020344.mm.1 |
TC0500002229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020344.mm.1 |
NM_001289803.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020344.mm.1 |
NM_001289804.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020344.mm.1 |
NM_181857.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020345.mm.1 |
TC0500002229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020345.mm.1 |
NM_001289803.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020345.mm.1 |
NM_001289804.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020345.mm.1 |
NM_181857.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020346.mm.1 |
TC0500002229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020346.mm.1 |
NM_001289804.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020346.mm.1 |
NM_181857.4 |
Refseq |