Assay Details
| Gene Symbol: Poln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289803.1 | NP_001276732.1 | 16 - 17 | 1957 | 89 | |
| NM_001289804.1 | NP_001276733.1 | 18 - 19 | 2060 | 89 | ||
| NM_181857.4 | NP_862905.1 | 17 - 18 | 2011 | 89 | ||
| GenBank mRNA | AK133063.1 | - | 18 - 19 | 1980 | 89 | |
| AY135562.3 | - | 17 - 18 | 1933 | 89 | ||
| BC141101.1 | - | 17 - 18 | 1911 | 9055713 | 89 | |
| BC145378.1 | - | 16 - 17 | 1857 | 9053936 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poln
Genomic Map