Assay Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290454.1 | NP_001277383.1 | 4 - 5 | 1322 | 84 | |
| GenBank mRNA | AK009596.1 | - | 4 - 5 | 1281 | 84 | |
More Information
Set Membership: |
|

Ubap1
Genomic Map