Assay Details
| Gene Symbol: Rars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181406.3 | NP_852071.2 | 5 - 6 | 428 | 136 | |
| GenBank mRNA | AK075721.1 | - | 5 - 6 | 409 | 136 | |
| AK156182.1 | - | 5 - 6 | 406 | 136 | ||
| AK166986.1 | - | 5 - 6 | 944 | 136 | ||
| BC024878.1 | - | 5 - 6 | 408 | 3989666 | 136 | |
| BU961647.1 | - | 430 | 136 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rars2
Genomic Map