Assay Details
| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175234.4 | NP_780443.2 | 2 - 3 | 632 | 95 | |
| GenBank mRNA | AK082464.1 | - | 2 - 3 | 626 | 95 | |
| AK144924.1 | - | 2 - 3 | 633 | 95 | ||
| BC059864.1 | - | 2 - 3 | 146 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059864.1 transcripts.
Set Membership: |
|

Faxc
Genomic Map