Assay Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 1 - 2 | 218 | 105 | |
| NM_001290420.1 | NP_001277349.1 | 1 - 2 | 218 | 105 | ||
| NM_001290422.1 | NP_001277351.1 | 1 - 2 | 218 | 105 | ||
| NM_016746.4 | NP_058026.2 | 1 - 2 | 218 | 105 | ||
| GenBank mRNA | AK009615.1 | - | 1 - 2 | 163 | 105 | |
| AK035728.1 | - | 1 - 2 | 163 | 105 | ||
| AK035762.1 | - | 1 - 2 | 179 | 105 | ||
| AK037957.1 | - | 1 - 2 | 219 | 105 | ||
| AK135472.1 | - | 1 - 2 | 214 | 105 | ||
| AK153035.1 | - | 1 - 2 | 151 | 105 | ||
| BB631264.1 | - | 1 - 2 | 216 | 105 | ||
| BC003344.1 | - | 1 - 2 | 151 | 3257730 | 105 | |
| BC029211.1 | - | 1 - 2 | 164 | 4526925 | 105 | |
| BC037689.1 | - | 1 - 2 | 146 | 105 | ||
| BC062376.1 | - | 1 - 2 | 133 | 105 | ||
| BC087544.1 | - | 1 - 2 | 150 | 105 | ||
| BC100396.1 | - | 1 - 2 | 192 | 105 | ||
| BC120649.1 | - | 1 - 2 | 145 | 40129572 | 105 | |
| BC120677.1 | - | 1 - 2 | 145 | 40129600 | 105 | |
| U62638.1 | - | 1 - 2 | 193 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037957.1, AK135472.1, AK153035.1, BC003344.1, BC029211.1, BC120649.1, BC120677.1, NM_001122982.2, NM_001290420.1, NM_001290422.1, NM_016746.4, U62638.1 transcripts.
Set Membership: |
|

Ccnc
Genomic Map