Assay Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 5 - 6 | 521 | 120 | |
| NM_001290420.1 | NP_001277349.1 | 5 - 6 | 521 | 120 | ||
| NM_001290422.1 | NP_001277351.1 | 5 - 6 | 521 | 120 | ||
| NM_016746.4 | NP_058026.2 | 5 - 6 | 521 | 120 | ||
| GenBank mRNA | AK014079.1 | - | 4 - 5 | 370 | 120 | |
| AK037957.1 | - | 5 - 6 | 522 | 120 | ||
| AK135472.1 | - | 5 - 6 | 517 | 120 | ||
| AK153035.1 | - | 5 - 6 | 454 | 120 | ||
| BB631264.1 | - | 5 - 6 | 519 | 120 | ||
| BC003344.1 | - | 5 - 6 | 454 | 3257730 | 120 | |
| BC037689.1 | - | 5 - 6 | 449 | 120 | ||
| BC062376.1 | - | 5 - 6 | 436 | 120 | ||
| BC087544.1 | - | 5 - 6 | 453 | 120 | ||
| BC100396.1 | - | 5 - 6 | 495 | 120 | ||
| BC120649.1 | - | 5 - 6 | 448 | 40129572 | 120 | |
| BC120677.1 | - | 5 - 6 | 448 | 40129600 | 120 | |
| U62638.1 | - | 5 - 6 | 496 | 120 | ||
More Information
Set Membership: |
|

Ccnc
Genomic Map