Assay Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 10 - 11 | 852 | 150 | |
| NM_001290420.1 | NP_001277349.1 | 10 - 11 | 852 | 150 | ||
| NM_001290422.1 | NP_001277351.1 | 10 - 11 | 852 | 150 | ||
| NM_016746.4 | NP_058026.2 | 10 - 11 | 852 | 150 | ||
| GenBank mRNA | AK009615.1 | - | 10 - 11 | 798 | 150 | |
| AK014079.1 | - | 9 - 10 | 701 | 150 | ||
| AK037957.1 | - | 10 - 11 | 853 | 150 | ||
| AK135472.1 | - | 10 - 11 | 848 | 150 | ||
| BC003344.1 | - | 10 - 11 | 785 | 3257730 | 150 | |
| BC029211.1 | - | 10 - 11 | 794 | 4526925 | 150 | |
| BC037689.1 | - | 10 - 11 | 780 | 150 | ||
| BC062376.1 | - | 10 - 11 | 767 | 150 | ||
| BC087544.1 | - | 10 - 11 | 784 | 150 | ||
| BC100396.1 | - | 10 - 11 | 826 | 150 | ||
| BC120649.1 | - | 10 - 11 | 779 | 40129572 | 150 | |
| BC120677.1 | - | 10 - 11 | 779 | 40129600 | 150 | |
| BQ175960.1 | - | 2 - 3 | 150 | |||
| U62638.1 | - | 10 - 11 | 827 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnc
Genomic Map