Assay Details
| Gene Symbol: Ccne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037134.2 | NP_001032211.1 | 4 - 5 | 387 | 72 | |
| NM_001282943.1 | NP_001269872.1 | 4 - 5 | 300 | 72 | ||
| NM_009830.3 | NP_033960.1 | 4 - 5 | 297 | 72 | ||
| GenBank mRNA | AF091432.1 | - | 3 - 4 | 192 | 72 | |
| AF106691.1 | - | 2 - 3 | 167 | 72 | ||
| AK010929.1 | - | 4 - 5 | 227 | 72 | ||
| AK031116.1 | - | 4 - 5 | 224 | 72 | ||
| AK037791.1 | - | 4 - 5 | 227 | 72 | ||
| AK158720.1 | - | 4 - 5 | 813 | 72 | ||
| AK167428.1 | - | 4 - 5 | 227 | 72 | ||
| BC053727.1 | - | 3 - 4 | 187 | 30061057 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccne2
Genomic Map