| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001048005.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001048005.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253777.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253777.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253778.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253778.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253780.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253780.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253788.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_001253788.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_028624.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NM_028624.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NR_045549.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NR_045549.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NR_045551.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
NR_045551.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400019943.mm.1 |
TC0400002470.mm.1 |
MTA TC |