Assay Details
Gene Symbol: 1110017D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048005.1 | NP_001041470.1 | 5 - 6 | 863 | 83 | |
NM_028624.1 | NP_082900.1 | 2 - 3 | 470 | 83 | ||
NR_045551.2 | 2 - 3 | 411 | 83 | |||
GenBank mRNA | AK003742.1 | - | 2 - 3 | 461 | 83 | |
BC049629.1 | - | 2 - 3 | 401 | 83 | ||
CA463601.1 | - | 2 - 3 | 473 | 83 | ||
DQ873295.1 | - | 2 - 3 | 332 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862464 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862460 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862466 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862452 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862465 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862528 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862459 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862467 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862462 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862463 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862527 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862469 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862458 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862457 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862461 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862455 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862529 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862470 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133365 | AB231294.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133365 | AK003742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133365 | NM_028624.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133365 | CA463601.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133365 | DQ873295.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069470 | AB231294.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069470 | AK003742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069470 | NM_028624.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069470 | BC049629.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069470 | BU700151.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116003 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603271 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309813 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393915 | AK079022.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393915 | AK153666.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32379 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47811 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5547 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150172 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069470 | AB231294.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069470 | AK003742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069470 | NM_028624.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069470 | BC049629.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069470 | BU700151.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133365 | AB231294.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133365 | AK003742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133365 | NM_028624.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133365 | CA463601.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133365 | DQ873295.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150172 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05212711 | - | - |
Whole Mouse Genome G4122A | A_51_P393567 | AB231294.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393567 | AK003742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393567 | NM_028624.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393567 | BC049629.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393567 | BU700151.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150172 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810983 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810985 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810987 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810980 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03654602 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810988 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810981 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03654598 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03810984 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862464 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862462 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862528 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862469 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862470 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862527 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862460 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862467 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862455 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862459 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862465 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862452 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862457 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862466 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862529 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862461 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862463 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22862458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799610 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04038798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862579 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03987578 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03480057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03839002 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03263048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03654601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03987572 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03810983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862540 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862586 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03945031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862565 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862555 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03261711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03810984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862559 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862588 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03309997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03248760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03248767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03489725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03810980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862595 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10512258 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512255 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512257 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512262 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512261 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512260 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512256 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512263 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512264 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512259 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429471_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10512254 | - | - |
Mouse Genome 430 2.0 Array | 1429471_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429471_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73721_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73721_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_001048005.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_001048005.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_001253777.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_001253777.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_001253778.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_001253778.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_028624.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NM_028624.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NR_045549.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NR_045549.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NR_045551.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | NR_045551.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019940.mm.1 | TC0400002470.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429471_PM_at | - | - |
Mouse Expression Set 430 | 1429471_at | - | - |
Murine Genome U74v2 Set | 165473_PM_at | - | - |
Murine Genome U74v2 Set | 165473_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429471_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429471_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 870136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6450746 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2617920 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2617920 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2617920 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3118081 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3043178 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3118081 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3043178 | - | - |
