Assay Details
| Gene Symbol: Rad54b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039556.3 | NP_001034645.1 | 2 - 3 | 242 | 101 | |
| NM_001256145.1 | NP_001243074.1 | 2 - 3 | 242 | 101 | ||
| GenBank mRNA | AK078439.1 | - | 2 - 3 | 220 | 101 | |
| BC057604.1 | - | 2 - 3 | 190 | 6416213 | 101 | |
| CJ160401.1 | - | 2 - 3 | 243 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078439.1 transcripts.
Set Membership: |
|

Rad54b
Genomic Map