Assay Details
| Gene Symbol: Rad54b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039556.3 | NP_001034645.1 | 7 - 8 | 1215 | 154 | |
| GenBank mRNA | BC057604.1 | - | 7 - 8 | 1163 | 6416213 | 154 |
More Information
Updated Mapping Information
Set Membership: |
|

Rad54b
Genomic Map