Assay Details
| Gene Symbol: Epha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122889.1 | NP_001116361.1 | 2 - 3 | 397 | 83 | |
| NM_001290434.1 | NP_001277363.1 | 2 - 3 | 411 | 83 | ||
| NM_010141.4 | NP_034271.3 | 2 - 3 | 411 | 83 | ||
| GenBank mRNA | AK030480.1 | - | 2 - 3 | 378 | 83 | |
| AK032973.1 | - | 2 - 3 | 216 | 83 | ||
| AK033903.1 | - | 2 - 3 | 412 | 83 | ||
| AK049848.1 | - | 2 - 3 | 378 | 83 | ||
| BC026153.1 | - | 2 - 3 | 303 | 3991628 | 83 | |
| CF729611.1 | - | 2 - 3 | 397 | 83 | ||
| X79082.1 | - | 2 - 3 | 390 | 83 | ||
| X79083.1 | - | 2 - 3 | 390 | 83 | ||
| X79084.1 | - | 2 - 3 | 390 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha7
Genomic Map