Assay Details
| Gene Symbol: Epha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290434.1 | NP_001277363.1 | 11 - 12 | 2355 | 85 | |
| NM_010141.4 | NP_034271.3 | 11 - 12 | 2367 | 85 | ||
| GenBank mRNA | AK030480.1 | - | 11 - 12 | 2334 | 85 | |
| BC026153.1 | - | 11 - 12 | 2247 | 3991628 | 85 | |
| X79082.1 | - | 11 - 12 | 2346 | 85 | ||
| X81466.1 | - | 7 - 8 | 824 | 85 | ||
More Information
Set Membership: |
|

Epha7
Genomic Map