Assay Details
| Gene Symbol: Epha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290434.1 | NP_001277363.1 | 15 - 16 | 2959 | 100 | |
| NM_010141.4 | NP_034271.3 | 15 - 16 | 2971 | 100 | ||
| GenBank mRNA | AK030480.1 | - | 15 - 16 | 2938 | 100 | |
| X79082.1 | - | 15 - 16 | 2950 | 100 | ||
| X81466.1 | - | 11 - 12 | 1428 | 100 | ||
More Information
Set Membership: |
|

Epha7
Genomic Map