Assay Details
Gene Symbol: Nkain3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290410.1 | NP_001277339.1 | 5 - 6 | 890 | 73 | |
GenBank mRNA | AK141456.1 | - | 5 - 6 | 790 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
