Assay Details
| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080927.2 | NP_001074396.1 | 6 - 7 | 855 | 113 | |
| NM_001080928.1 | NP_001074397.1 | 6 - 7 | 741 | 113 | ||
| NM_001277116.1 | NP_001264045.1 | 8 - 9 | 937 | 113 | ||
| NM_009035.5 | NP_033061.3 | 6 - 7 | 757 | 113 | ||
| GenBank mRNA | AK028363.1 | - | 8 - 9 | 915 | 113 | |
| AK080359.1 | - | 6 - 7 | 650 | 113 | ||
| AK090006.1 | - | 6 - 7 | 642 | 112 | ||
| AK139087.1 | - | 6 - 7 | 642 | 113 | ||
| AK145187.1 | - | 6 - 7 | 649 | 113 | ||
| AK152746.1 | - | 6 - 7 | 757 | 113 | ||
| AK153163.1 | - | 7 - 8 | 645 | 113 | ||
| BC035299.1 | - | 4 - 5 | 543 | 113 | ||
| BC051387.1 | - | 6 - 7 | 686 | 4460290 | 113 | |
| S63463.1 | - | 6 - 7 | 758 | 113 | ||
| X17459.1 | - | 6 - 7 | 753 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbpj
Genomic Map