Assay Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023047.2 | NP_075534.1 | 5 - 6 | 952 | 64 | |
| GenBank mRNA | AF249295.1 | - | 5 - 6 | 1116 | 64 | |
| AK082132.1 | - | 2 - 3 | 98 | 64 | ||
| BC065054.1 | - | 5 - 6 | 952 | 5709296 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl5
Genomic Map